4,4',4''-(Pyridine-2,4,6-triyl)tribenzaldehyde 98%

Supplier: Ambeed
Warning

A1164710-100MG A1164710-1G A1164710-250MG
76675-676EA 12 USD
76675-676 76675-678 76675-680
4,4',4''-(Pyridine-2,4,6-triyl)tribenzaldehyde 98%
4,4',4''-(Pyridine-2,4,6-triyl)tribenzaldehyde

Precaución: for Research Use Only

Formula: C₂₆H₁₇NO₃
CAS Number: 2230887-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR