[1-[(3,5-Dimethyl-1H-pyrrol-2-yl)(3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]-4-iodobenzene](difluoroborane) 98%

Supplier: Ambeed
Warning

Synonyms: Boron, [2-[(3,5-dimethyl-2H-pyrrol-2-ylidene-κN)(4-iodophenyl)methyl]-3,5-dimethyl-1H-pyrrolato-κN]difluoro-, (T-4)-

A1164555-100MG A1164555-1G A1164555-250MG
76675-670EA 66.84 USD
76675-670 76675-672 76675-674
[1-[(3,5-Dimethyl-1H-pyrrol-2-yl)(3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]-4-iodobenzene](difluoroborane) 98%
[1-[(3,5-Dimethyl-1H-pyrrol-2-yl)(3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]-4-iodobenzene](difluoroborane)

Precaución: for Research Use Only

Formula: C₁₉H₁₈BF₂IN₂
MW: 450.08 g/mol
MDL Number: MFCD29905036
CAS Number: 250734-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR