Copper(II) bis(trifluoromethanesulfonyl)imide 98%

Supplier: Ambeed
Danger

Synonyms: Bis(trifluoromethanesulphonyl)imide Copper(II) salt, Copper(II) triflimide, Copper(II) bis(trifluoromethylsulphonyl)imide

A1164533-1G A1164533-250MG A1164533-25G A1164533-5G
76675-662EA 41 USD
76675-662 76675-664 76675-666 76675-668
Copper(II) bis(trifluoromethanesulfonyl)imide 98%
Copper(II) bis(trifluoromethanesulfonyl)imide

Precaución: for Research Use Only

Formula: C₄CuF₁₂N₂O₈S₄
MW: 623.82 g/mol
CAS Number: 162715-14-2
UN: 1759
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR