1,3,5,7-tetrakis(4-formylphenyl)adamantane 98%

Supplier: Ambeed
Warning

A1164189-100MG A1164189-50MG A1164189-250MG
76675-658EA 123.07 USD
76675-658 76675-660 77620-336
1,3,5,7-tetrakis(4-formylphenyl)adamantane 98%
1,3,5,7-tetrakis(4-formylphenyl)adamantane

Precaución: for Research Use Only

Formula: C₃₈H₃₂O₄
MW: 552.6583 g/mol
CAS Number: 645401-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR