Bis([1,10-Phenanthroline)(4,5,9,14-Tetraaza-benzo[b]triphenylene)ruthenium (II) dichloride 98%

Supplier: Ambeed
Warning

Synonyms: (OC-6-22-Delta)-Bis(1,10-phenanthroline)(dipyridophenazine)ruthenium dichloride

A1163949-100MG A1163949-1G A1163949-250MG
76675-652EA 143 USD
76675-652 76675-654 76675-656
Bis([1,10-Phenanthroline)(4,5,9,14-Tetraaza-benzo[b]triphenylene)ruthenium (II) dichloride 98%
Bis([1,10-Phenanthroline)(4,5,9,14-Tetraaza-benzo[b]triphenylene)ruthenium (II) dichloride

Precaución: for Research Use Only

Formula: C₄₂H₂₆Cl₂N₈Ru
MW: 814.68 g/mol
CAS Number: 132045-66-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR