N2-(N-glycyl-L-histidyl)-L-Lysine, copper complex 99%

Supplier: Ambeed
Warning

Synonyms: GHK-Cu, Copper tripeptide

A1163903-1G A1163903-250MG A1163903-5G
76675-646EA 39.26 USD
76675-646 76675-648 76675-650
N2-(N-glycyl-L-histidyl)-L-Lysine, copper complex 99%
N2-(N-glycyl-L-histidyl)-L-Lysine, copper complex

Precaución: for Research Use Only

Formula: C₁₄H₂₂CuN₆O₄
MW: 401.9083 g/mol
MDL Number: MFCD09842093
CAS Number: 89030-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR