(4-((1-Methylpiperidin-4-yl)oxy)phenyl)methanol 97%

Supplier: Ambeed
Warning

A1163871-100MG A1163871-1G A1163871-250MG A1163871-25G A1163871-5G
76675-636EA 46.68 USD
76675-636 76675-638 76675-640 76675-642 76675-644
(4-((1-Methylpiperidin-4-yl)oxy)phenyl)methanol 97%
(4-((1-Methylpiperidin-4-yl)oxy)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO₂
MW: 221.3 g/mol
CAS Number: 1782394-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR