2,2-Dimethyl-4-oxo-3,8,11,14,17,20,23,26,29-nonaoxa-5-azadotriacontan-32-oic acid 95%

Supplier: Ambeed
Warning

Synonyms: Boc-NH-PEG8-CH2CH2COOH

A1163457-100MG A1163457-1G A1163457-250MG A1163457-5G A1163457-25G
76675-628EA 59.42 USD
76675-628 76675-630 76675-632 76675-634 77620-322
2,2-Dimethyl-4-oxo-3,8,11,14,17,20,23,26,29-nonaoxa-5-azadotriacontan-32-oic acid 95%
2,2-Dimethyl-4-oxo-3,8,11,14,17,20,23,26,29-nonaoxa-5-azadotriacontan-32-oic acid

Precaución: for Research Use Only

Formula: C₂₄H₄₇NO₁₂
MW: 541.62 g/mol
MDL Number: MFCD13185254
CAS Number: 1334169-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR