N-(4-Chlorophenyl)-3-((4-chlorophenyl)amino)-2-(3-methylisoxazol-5-yl)acrylamide 97%
Supplier: Ambeed
|
Warning
|
A1163381-10MG
A1163381-1MG
A1163381-50MG
A1163381-5MG
76675-614EA
73.01
USD
76675-614
76675-616
76675-618
76675-620
N-(4-Chlorophenyl)-3-((4-chlorophenyl)amino)-2-(3-methylisoxazol-5-yl)acrylamide 97%
N-(4-Chlorophenyl)-3-((4-chlorophenyl)amino)-2-(3-methylisoxazol-5-yl)acrylamide
Precaución: for Research Use Only
Formula:
C₁₉H₁₅Cl₂N₃O₂ |
MDL Number:
MFCD18086897 CAS Number: 917837-54-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...