(4-(((tert-Butoxycarbonyl)amino)methyl)-3-fluorophenyl)boronic acid 98%

Supplier: Ambeed
Warning

A1163128-100MG A1163128-1G A1163128-250MG A1163128-5G
76675-606EA 41.38 USD
76675-606 76675-608 76675-610 76675-612
(4-(((tert-Butoxycarbonyl)amino)methyl)-3-fluorophenyl)boronic acid 98%
(4-(((tert-Butoxycarbonyl)amino)methyl)-3-fluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₇BFNO₄
MW: 269.0771 g/mol
MDL Number: MFCD29086649
CAS Number: 1374451-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR