Methyl 4-amino-5-bromo-2-chlorobenzoate 98%

Supplier: Ambeed
Warning

A1163077-1G A1163077-250MG A1163077-5G
76675-600EA 57.29 USD
76675-600 76675-602 76675-604
Methyl 4-amino-5-bromo-2-chlorobenzoate 98%
Methyl 4-amino-5-bromo-2-chlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₇BrClNO₂
MW: 264.5037 g/mol
MDL Number: MFCD23707492
CAS Number: 868406-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR