(2-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)(pyrrolidin-1-yl)methanone 98%

Supplier: Ambeed
Warning

A1162007-100MG A1162007-1G A1162007-250MG
76675-586EA 37 USD
76675-586 76675-588 76675-590
(2-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)(pyrrolidin-1-yl)methanone 98%
(2-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)(pyrrolidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₈H₂₆BNO₃
MDL Number: MFCD26399448
CAS Number: 1092563-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR