N-(2-Hydroxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulfonamide 98%

Supplier: Ambeed
Warning

A1161960-100MG A1161960-1G A1161960-250MG
76675-580EA 58.35 USD
76675-580 76675-582 76675-584
N-(2-Hydroxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulfonamide 98%
N-(2-Hydroxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₄H₂₂BNO₅S
MW: 327.2042 g/mol
MDL Number: MFCD28397687
CAS Number: 1014613-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR