tert-Butyl (2-(2-iodoethoxy)ethyl)carbamate 95%

Supplier: Ambeed
Danger

A1161790-1G A1161790-250MG A1161790-5G A1161790-25G
76675-574EA 218 USD
76675-574 76675-576 76675-578 77620-312
tert-Butyl (2-(2-iodoethoxy)ethyl)carbamate 95%
tert-Butyl (2-(2-iodoethoxy)ethyl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₈INO₃
MDL Number: MFCD24467264
CAS Number: 629626-40-0
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR