4-(Cyclopentylmethyl)aniline 95%

Supplier: Ambeed
Warning

A1161734-100MG A1161734-1G A1161734-250MG
76675-568EA 103 USD
76675-568 76675-570 76675-572
4-(Cyclopentylmethyl)aniline 95%
4-(Cyclopentylmethyl)aniline

Precaución: for Research Use Only

Formula: C₁₂H₁₇N
MDL Number: MFCD20716142
CAS Number: 145071-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR