Sulindac sulfone 97%

Supplier: Ambeed
Warning

Synonyms: Z-5-Fluoro-2-methyl-1-[p-(methyl-sulphonyl)benzylidene]indene-3-acetic acid

A1161164-1MG A1161164-25MG A1161164-50MG
76675-558EA 10.61 USD
76675-558 76675-560 76675-562
Sulindac sulfone 97%
Sulindac sulfone

Precaución: for Research Use Only

Formula: C₂₀H₁₇FO₄S
MW: 372.4 g/mol
MDL Number: MFCD00871143
CAS Number: 59864-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR