2,5-Dioxopyrrolidin-1-yl hept-6-ynoate 95+%

Supplier: Ambeed
Warning

A1161129-250MG A1161129-5G A1161129-100MG A1161129-1G
76675-554EA 37.14 USD
76675-554 76675-556 77620-308 77620-310
2,5-Dioxopyrrolidin-1-yl hept-6-ynoate 95+%
2,5-Dioxopyrrolidin-1-yl hept-6-ynoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₄
MW: 223.2252 g/mol
MDL Number: MFCD29041721
CAS Number: 917222-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR