O-(2-(Prop-2-yn-1-yloxy)ethyl)hydroxylamine 98%

Supplier: Ambeed
Danger

Synonyms: Aminooxy-PEG1-propargyl

A1159765-100MG A1159765-1G A1159765-250MG
76675-544EA 49.87 USD
76675-544 77595-960 77595-962
O-(2-(Prop-2-yn-1-yloxy)ethyl)hydroxylamine 98%
O-(2-(Prop-2-yn-1-yloxy)ethyl)hydroxylamine

Precaución: for Research Use Only

Formula: C₅H₉NO₂
MW: 115.1305 g/mol
MDL Number: MFCD29764324
CAS Number: 1895922-69-6
UN: 2735
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR