4-Chloro-N-(2,5-dimethylphenyl)benzenesulfonamide 98%

Supplier: Ambeed
Warning

A1159449-1MG A1159449-25MG A1159449-50MG
76675-526EA 5 USD
76675-526 76675-528 76675-530
4-Chloro-N-(2,5-dimethylphenyl)benzenesulfonamide 98%
4-Chloro-N-(2,5-dimethylphenyl)benzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₄H₁₄ClNO₂S
MDL Number: MFCD00784288
CAS Number: 7454-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR