(S)-N-(2-Chloro-6-fluorophenyl)-4-(4-ethyl-3-(hydroxymethyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl)-5-fluoro-2-((1,1,1-trifluoropropan-2-yl)oxy)benzamide 97%

Supplier: Ambeed
Warning

Synonyms: BAY-2402234

A1159404-10MG A1159404-1MG A1159404-5MG
76675-508EA 166.57 USD
76675-508 76675-510 76675-512
(S)-N-(2-Chloro-6-fluorophenyl)-4-(4-ethyl-3-(hydroxymethyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl)-5-fluoro-2-((1,1,1-trifluoropropan-2-yl)oxy)benzamide 97%
(S)-N-(2-Chloro-6-fluorophenyl)-4-(4-ethyl-3-(hydroxymethyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl)-5-fluoro-2-((1,1,1-trifluoropropan-2-yl)oxy)benzamide

Precaución: for Research Use Only

Formula: C₂₁H₁₈ClF₅N₄O₄
MW: 520.84 g/mol
MDL Number: MFCD31692414
CAS Number: 2225819-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR