3,3-Dimethylindolin-4-amine dihydrochloride 97%

Supplier: Ambeed
Warning

A1159400-100MG A1159400-250MG
76675-504EA 214 USD
76675-504 76675-506
3,3-Dimethylindolin-4-amine dihydrochloride 97%
3,3-Dimethylindolin-4-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₆Cl₂N₂
CAS Number: 2411639-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR