(4-([2,2':6',2''-Terpyridin]-4'-yl)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A1159368-10G A1159368-1G A1159368-5G A1159368-250MG
76675-498EA 740 USD
76675-498 76675-500 76675-502 77595-952
(4-([2,2':6',2''-Terpyridin]-4'-yl)phenyl)boronic acid 97%
(4-([2,2':6',2''-Terpyridin]-4'-yl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₂₁H₁₆BN₃O₂
MW: 353.1816 g/mol
CAS Number: 381218-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR