Tetrabutylammonium chloride 95% < 7% H₂O

Supplier: Ambeed
Warning

A1159021-100G A1159021-25G A1159021-500G
76675-458EA 31 USD
76675-458 76675-460 76675-462
Tetrabutylammonium chloride 95% < 7% H₂O
Tetrabutylammonium chloride

Precaución: for Research Use Only

Formula: C₁₆H₃₆ClN
MDL Number: MFCD00011635
CAS Number: 1112-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR