2-Butynoyl chloride 97%

Supplier: Ambeed
Danger

A1158877-1G A1158877-250MG A1158877-100MG A1158877-5G
76675-450EA 145 USD
76675-450 77549-300 77866-121 77866-122
2-Butynoyl chloride 97%
2-Butynoyl chloride

Precaución: for Research Use Only

Formula: C₄H₃ClO
MDL Number: MFCD27950345
CAS Number: 39753-54-3
UN: 2927
ADR: 6.1,8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR