(R,R)-2,2'-Bis[(R)-(N,N-dimethylamino)(phenyl)methyl]-1,1'-bis(diphenylphosphino)ferrocene 98%
Supplier: Ambeed
|
Warning
|
A1158824-100MG
A1158824-250MG
A1158824-50MG
76675-444EA
76.39
USD
76675-444
76675-446
76675-448
(R,R)-2,2'-Bis[(R)-(N,N-dimethylamino)(phenyl)methyl]-1,1'-bis(diphenylphosphino)ferrocene 98%
(R,R)-2,2'-Bis[(R)-(N,N-dimethylamino)(phenyl)methyl]-1,1'-bis(diphenylphosphino)ferrocene
Precaución: for Research Use Only
Formula:
C₅₂H₅₀FeN₂P₂ MW: 820.76 g/mol |
MDL Number:
MFCD04117705 CAS Number: 174467-31-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...