cis-Dichlorobis(pyridine)platinum(II) 98%

Supplier: Ambeed
Warning

A1158813-100MG A1158813-1G A1158813-250MG
76675-438EA 24 USD
76675-438 76675-440 76675-442
cis-Dichlorobis(pyridine)platinum(II) 98%
cis-Dichlorobis(pyridine)platinum(II)

Precaución: for Research Use Only

Formula: C₁₀H₁₀Cl₂N₂Pt
MDL Number: MFCD00058858
CAS Number: 15227-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR