Bis(acetonitrile)(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate 98%

Supplier: Ambeed
Warning

Synonyms: Bis(acetonitrile)(1,5-cyclooctadiene)rhodium(I)tetrafluoroborate

A1158806-100MG A1158806-1G A1158806-250MG
76675-432EA 90.18 USD
76675-432 76675-434 76675-436
Bis(acetonitrile)(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate 98%
Bis(acetonitrile)(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂Rh
MW: 293.19 g/mol
MDL Number: MFCD06798085
CAS Number: 32679-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR