(R)-(+)-[(R)-2-Dicyclohexylphosphinoferrocenyl](N,N-dimethylamino)(2-dicyclohexylphosphinophenyl)methane 98%

Supplier: Ambeed
Warning

Synonyms: (1Rp)-1-(Dicyclohexylphosphino)-2-[(R)-[2-(dicyclohexylphosphino)phenyl](dimethylamino)methyl]ferrocene

A1158797-100MG A1158797-50MG A1158797-250MG A1158797-1G
76675-428EA 88 USD
76675-428 76675-430 77595-942 77866-119
(R)-(+)-[(R)-2-Dicyclohexylphosphinoferrocenyl](N,N-dimethylamino)(2-dicyclohexylphosphinophenyl)methane 98%
(R)-(+)-[(R)-2-Dicyclohexylphosphinoferrocenyl](N,N-dimethylamino)(2-dicyclohexylphosphinophenyl)methane

Precaución: for Research Use Only

Formula: C₄₃H₆₃FeNP₂
MW: 711.76 g/mol
MDL Number: MFCD04117706
CAS Number: 1156547-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR