(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine ≥98%, ee 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-2,2',6,6'-Tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine, CTH-(R)-P-Phos
A1158795-100MG
A1158795-1G
A1158795-250MG
A1158795-50MG
76675-420EA
120.95
USD
76675-420
76675-422
76675-424
76675-426
(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine ≥98%, ee 99%
(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine
Precaución: for Research Use Only
Formula:
C₃₈H₃₄N₂O₄P₂ MW: 644.64 g/mol |
MDL Number:
MFCD04038734 CAS Number: 221012-82-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...