(S,S)-1,2-Bis(t-butylmethylphosphino)benzene (S,S)-BenzP 98%

Supplier: Ambeed
Warning

A1158793-100MG A1158793-50MG A1158793-250MG
76675-416EA 176.11 USD
76675-416 76675-418 77570-970
(S,S)-1,2-Bis(t-butylmethylphosphino)benzene (S,S)-BenzP 98%
(S,S)-1,2-Bis(t-butylmethylphosphino)benzene (S,S)-BenzP

Precaución: for Research Use Only

Formula: C₁₆H₂₈P₂
MW: 282.3400 g/mol
MDL Number: MFCD18837015
CAS Number: 1355162-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR