4-Amino-N-(1-((3-chloro-2-fluorophenyl)amino)-6-methylisoquinolin-5-yl)thieno[3,2-d]pyrimidine-7-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: Belvarafenib

A1158776-10MG A1158776-1MG A1158776-5MG
76675-410EA 109.28 USD
76675-410 76675-412 76675-414
4-Amino-N-(1-((3-chloro-2-fluorophenyl)amino)-6-methylisoquinolin-5-yl)thieno[3,2-d]pyrimidine-7-carboxamide 98%
4-Amino-N-(1-((3-chloro-2-fluorophenyl)amino)-6-methylisoquinolin-5-yl)thieno[3,2-d]pyrimidine-7-carboxamide

Precaución: for Research Use Only

Formula: C₂₃H₁₆ClFN₆OS
MW: 478.93 g/mol
MDL Number: MFCD31731098
CAS Number: 1446113-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR