Ir[p-F(t-Bu)-ppy]3 98%

Supplier: Ambeed
Warning

A1158735-100MG A1158735-1G A1158735-250MG A1158735-50MG
76675-398EA 182 USD
76675-398 76675-400 76675-402 76675-404
Ir[p-F(t-Bu)-ppy]3 98%
Ir[p-F(t-Bu)-ppy]3

Precaución: for Research Use Only

Formula: C₄₅H₄₅F₃IrN₃
MDL Number: MFCD30737992
CAS Number: 1311386-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR