Ir[dF(t-Bu)-ppy]3 95%

Supplier: Ambeed
Warning

A1158734-100MG A1158734-1G A1158734-250MG
76675-394EA 132.62 USD
76675-394 76675-396 77549-298
Ir[dF(t-Bu)-ppy]3 95%
Ir[dF(t-Bu)-ppy]3

Precaución: for Research Use Only

Formula: C₄₅H₄₂F₆IrN₃
MW: 931.0425 g/mol
MDL Number: MFCD30737993
CAS Number: 1380203-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR