2,7-Naphthalenebis(trifluoromethanesulfonate) 97%

Supplier: Ambeed
Danger

Synonyms: 2,7-Bis(trifluoromethanesulphonyloxy)naphthalene, 2,7-Naphthaleneditriflate, Naphthalene-2,7-diyl bis(trifluoromethanesulphonate)

A1158683-1G A1158683-250MG A1158683-5G
76675-386EA 58.35 USD
76675-386 76675-388 76675-390
2,7-Naphthalenebis(trifluoromethanesulfonate) 97%
2,7-Naphthalenebis(trifluoromethanesulfonate)

Precaución: for Research Use Only

Formula: C₁₂H₆F₆O₆S₂
MW: 424.28 g/mol
MDL Number: MFCD20260329
CAS Number: 151391-00-3
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR