(2,4,6-Triethylbenzene-1,3,5-triyl)trimethanamine trihydrochloride 95%
Supplier: Ambeed
|
Warning
|
A1158523-100MG
A1158523-1G
A1158523-250MG
76675-368EA
43
USD
76675-368
76675-370
76675-372
(2,4,6-Triethylbenzene-1,3,5-triyl)trimethanamine trihydrochloride 95%
(2,4,6-Triethylbenzene-1,3,5-triyl)trimethanamine trihydrochloride
Precaución: for Research Use Only
Formula:
C₁₅H₃₀Cl₃N₃ |
MDL Number:
MFCD15144745 CAS Number: 190779-64-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...