Benzenemethanamine, 4-[4-[4-(trifluoromethoxy)phenyl]-1-piperidinyl]- 98%

Supplier: Ambeed
Warning

A1158045-250MG A1158045-5G
76675-348EA 35.01 USD
76675-348 76675-350
Benzenemethanamine, 4-[4-[4-(trifluoromethoxy)phenyl]-1-piperidinyl]- 98%
Benzenemethanamine, 4-[4-[4-(trifluoromethoxy)phenyl]-1-piperidinyl]-

Precaución: for Research Use Only

Formula: C₁₉H₂₁F₃N₂O
MW: 350.3781 g/mol
MDL Number: MFCD29033786
CAS Number: 1426520-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR