Boron, [N-[(carboxy-κO)methyl]-N-methylglycinato(2-)-κN,κO][4-(hydroxymethyl)phenyl]-, (T-4)- 98%

Supplier: Ambeed
Warning

A1158031-1G A1158031-25G A1158031-5G
76675-342EA 46.68 USD
76675-342 76675-344 76675-346
Boron, [N-[(carboxy-κO)methyl]-N-methylglycinato(2-)-κN,κO][4-(hydroxymethyl)phenyl]-, (T-4)- 98%
Boron, [N-[(carboxy-κO)methyl]-N-methylglycinato(2-)-κN,κO][4-(hydroxymethyl)phenyl]-, (T-4)-

Precaución: for Research Use Only

Formula: C₁₂H₁₄BNO₅
MW: 263.0543 g/mol
MDL Number: MFCD11215229
CAS Number: 1072960-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR