tert-Butyl 5-oxo-5,7-dihydrospiro[cyclopenta[b]pyridine-6,4'-piperidine]-1'-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A1157504-100MG
A1157504-1G
A1157504-250MG
76675-324EA
78.51
USD
76675-324
76675-326
76675-328
tert-Butyl 5-oxo-5,7-dihydrospiro[cyclopenta[b]pyridine-6,4'-piperidine]-1'-carboxylate 98%
tert-Butyl 5-oxo-5,7-dihydrospiro[cyclopenta[b]pyridine-6,4'-piperidine]-1'-carboxylate
Precaución: for Research Use Only
Formula:
C₁₇H₂₂N₂O₃ MW: 302.3682 g/mol |
MDL Number:
MFCD32173167 CAS Number: 2245084-40-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...