5-Bromo-4-fluoropyridin-3-ol 96%

Supplier: Ambeed
Warning

A1157116-1G A1157116-100MG A1157116-250MG
76675-308EA 499 USD
76675-308 77866-104 77866-105
5-Bromo-4-fluoropyridin-3-ol 96%
5-Bromo-4-fluoropyridin-3-ol

Precaución: for Research Use Only

Formula: C₅H₃BrFNO
MDL Number: MFCD26517020
CAS Number: 1805515-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR