3-Iodo-1-tosyl-1H-indole 97%

Supplier: Ambeed
Warning

A1157113-100MG A1157113-1G A1157113-250MG A1157113-5G
76675-300EA 12 USD
76675-300 76675-302 76675-304 76675-306
3-Iodo-1-tosyl-1H-indole 97%
3-Iodo-1-tosyl-1H-indole

Precaución: for Research Use Only

Formula: C₁₅H₁₂INO₂S
MDL Number: MFCD13177490
CAS Number: 170456-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR