2'-Bromospiro[fluorene-9,9'-xanthene] 98%

Supplier: Ambeed
Warning

A1157088-1G A1157088-5G
76675-296EA 28 USD
76675-296 76675-298
2'-Bromospiro[fluorene-9,9'-xanthene] 98%
2'-Bromospiro[fluorene-9,9'-xanthene]

Precaución: for Research Use Only

Formula: C₂₅H₁₅BrO
MDL Number: MFCD31814409
CAS Number: 1477458-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR