9-(4-(4-Bromophenyl)-6-phenyl-1,3,5-triazin-2-yl)-9H-carbazole 98%

Supplier: Ambeed
Warning

A1157077-1G A1157077-250MG
76675-288EA 55.17 USD
76675-288 76675-290
9-(4-(4-Bromophenyl)-6-phenyl-1,3,5-triazin-2-yl)-9H-carbazole 98%
9-(4-(4-Bromophenyl)-6-phenyl-1,3,5-triazin-2-yl)-9H-carbazole

Precaución: for Research Use Only

Formula: C₂₇H₁₇BrN₄
MW: 477.3547 g/mol
MDL Number: MFCD32206907
CAS Number: 1616413-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR