Methyl 2-amino-4-chlorobenzo[d]thiazole-6-carboxylate 97%

Supplier: Ambeed
Warning

A1157032-100MG A1157032-250MG
76675-282EA 48.81 USD
76675-282 76675-284
Methyl 2-amino-4-chlorobenzo[d]thiazole-6-carboxylate 97%
Methyl 2-amino-4-chlorobenzo[d]thiazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇ClN₂O₂S
MW: 242.6821 g/mol
MDL Number: MFCD23705750
CAS Number: 1427425-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR