4-(((4-(4-(2-Chlorophenyl)piperazin-1-yl)-6-phenylpyrimidin-2-yl)thio)methyl)benzoic acid 99%

Supplier: Ambeed
Warning

Synonyms: J14

A1157030-10MG A1157030-1MG A1157030-25MG A1157030-50MG A1157030-5MG
76675-272EA 88.06 USD
76675-272 76675-274 76675-276 76675-278 76675-280
4-(((4-(4-(2-Chlorophenyl)piperazin-1-yl)-6-phenylpyrimidin-2-yl)thio)methyl)benzoic acid 99%
4-(((4-(4-(2-Chlorophenyl)piperazin-1-yl)-6-phenylpyrimidin-2-yl)thio)methyl)benzoic acid

Precaución: for Research Use Only

Formula: C₂₈H₂₅ClN₄O₂S
MW: 517.0417 g/mol
MDL Number: MFCD19814303
CAS Number: 1043854-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR