4,4',4'',4'''-(Pyrene-1,3,6,8-tetrayltetrakis(ethyne-2,1-diyl))tetrabenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A1156895-100MG
A1156895-250MG
76675-266EA
62.6
USD
76675-266
76675-268
4,4',4'',4'''-(Pyrene-1,3,6,8-tetrayltetrakis(ethyne-2,1-diyl))tetrabenzoic acid 98%
4,4',4'',4'''-(Pyrene-1,3,6,8-tetrayltetrakis(ethyne-2,1-diyl))tetrabenzoic acid
Precaución: for Research Use Only
Formula:
C₅₂H₂₆O₈ MW: 778.7581 g/mol |
CAS Number:
1569900-71-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...