3,7-Dibenzothiophenedicarboxylic acid, 5,5-dioxide 98%

Supplier: Ambeed
Warning

A1156894-1G A1156894-250MG A1156894-100MG
76675-264EA 56 USD
76675-264 77866-100 77866-099
3,7-Dibenzothiophenedicarboxylic acid, 5,5-dioxide 98%
3,7-Dibenzothiophenedicarboxylic acid, 5,5-dioxide

Precaución: for Research Use Only

Formula: C₁₄H₈O₆S
MDL Number: MFCD00413883
CAS Number: 23613-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR