4,4'-Oxydiphthalic acid 98%

Supplier: Ambeed
Warning

A1156888-100MG A1156888-1G A1156888-250MG A1156888-5G
76675-256EA 24.41 USD
76675-256 76675-258 76675-260 76675-262
4,4'-Oxydiphthalic acid 98%
4,4'-Oxydiphthalic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₀O₉
MW: 346.2452 g/mol
MDL Number: MFCD00378118
CAS Number: 7717-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR