1,4-Bis(dodecyloxy)-2,5-diethynylbenzene 98%

Supplier: Ambeed
Warning

A1156881-100MG A1156881-1G A1156881-250MG A1156881-5G
76675-240EA 29.71 USD
76675-240 76675-242 76675-244 76675-246
1,4-Bis(dodecyloxy)-2,5-diethynylbenzene 98%
1,4-Bis(dodecyloxy)-2,5-diethynylbenzene

Precaución: for Research Use Only

Formula: C₃₄H₅₄O₂
MW: 494.7914 g/mol
CAS Number: 152270-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR