1,4-Bis(pyridin-4-ylethynyl)benzene 98%

Supplier: Ambeed
Warning

A1156878-100MG A1156878-250MG A1156878-5G
76675-226EA 47.75 USD
76675-226 76675-228 76675-230
1,4-Bis(pyridin-4-ylethynyl)benzene 98%
1,4-Bis(pyridin-4-ylethynyl)benzene

Precaución: for Research Use Only

Formula: C₂₀H₁₂N₂
MW: 280.3200 g/mol
CAS Number: 158525-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR