1,3-Di(1H-imidazol-1-yl)benzene 98%

Supplier: Ambeed
Warning

A1156877-100MG A1156877-1G A1156877-250MG A1156877-5G
76675-218EA 20.16 USD
76675-218 76675-220 76675-222 76675-224
1,3-Di(1H-imidazol-1-yl)benzene 98%
1,3-Di(1H-imidazol-1-yl)benzene

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₄
MW: 210.2346 g/mol
MDL Number: MFCD23103199
CAS Number: 69506-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR